Anti-NFE2L2 Rabbit Monoclonal Antibody [clone: EP1808Y]
Leverandør: Abcam
ABCAAB62352-100EA 12900 NOK
ABCAAB62352-100
Anti-NFE2L2 Rabbit Monoclonal Antibody [clone: EP1808Y]
Antistoffer
Type: Primary
Antigen: NFE2L2 (nuclear factor, erythroid 2-like 2)
Clonality: monoclonal
Clone: EP1808Y
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå