Anti-PDGFRB Rabbit Monoclonal Antibody [clone: EP2136Y]

Leverandør: Abcam
ABCAAB62367-100EA 10625 NOK
ABCAAB62367-100
Anti-PDGFRB Rabbit Monoclonal Antibody [clone: EP2136Y]
Antistoffer
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: monoclonal
Clone: EP2136Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR