Anti-PTPN11 Rabbit Monoclonal Antibody [clone: EP509Y]
Leverandør: Abcam
ABCAAB62379-100EA 11425 NOK
ABCAAB62379-100
Anti-PTPN11 Rabbit Monoclonal Antibody [clone: EP509Y]
Antistoffer
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: monoclonal
Clone: EP509Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå