Anti-SCN5A Mouse Monoclonal Antibody [clone: 4G8:1G7]
Leverandør: Abcam
ABCAAB62388-100EA 13525 NOK
ABCAAB62388-100
Anti-SCN5A Mouse Monoclonal Antibody [clone: 4G8:1G7]
Antistoffer
Type: Primary
Antigen: SCN5A (sodium channel, voltage-gated, type V, alpha subunit)
Clonality: monoclonal
Clone: 4G8:1G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human