Anti-TRAF6 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB62488-100EA 11775 NOK
ABCAAB62488-100
Anti-TRAF6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRAF6 (TNF receptor-associated factor 6, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat