Anti-Avian Influenza A Hemagglutinin Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB62490-100EA 11400 NOK
ABCAAB62490-100
Anti-Avian Influenza A Hemagglutinin Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Avian Influenza A Hemagglutinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR