Anti-Mineralocorticoid Receptor Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62532-100EA 8425 NOK
ABCAAB62532-100
Anti-Mineralocorticoid Receptor Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Mineralocorticoid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR