Anti-C12orf5 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62533-50EA 11775 NOK
ABCAAB62533-50
Anti-C12orf5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR