Anti-MICA Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB62540-100EA 11725 NOK
ABCAAB62540-100
Anti-MICA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MICA (MHC class I polypeptide-related sequence A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR