Anti-ADIPOQ Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB62551-100EA 11725 NOK
ABCAAB62551-100
Anti-ADIPOQ Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat