Anti-BECN1 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB62557-50EA 9000 NOK
ABCAAB62557-50
Anti-BECN1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BECN1 (beclin 1, autophagy related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR