Anti-ATF6 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB62576-100EA 11775 NOK
ABCAAB62576-100
Anti-ATF6 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR