Anti-SOCS1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB62584-100EA 12000 NOK
ABCAAB62584-100
Anti-SOCS1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat