Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]
Leverandør: Abcam
ABCAAB62623-100EA 12275 NOK
ABCAAB62623-100
Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]
Antistoffer
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: monoclonal
Clone: 15A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå