Anti-MBP Mouse Monoclonal Antibody [clone: MBP101]
Leverandør: Abcam
ABCAAB62631-100EA 12175 NOK
ABCAAB62631-100
Anti-MBP Mouse Monoclonal Antibody [clone: MBP101]
Antistoffer
Type: Primary
Antigen: MBP (myelin basic protein)
Clonality: monoclonal
Clone: MBP101
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå