Anti-D-glucosamine Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB62666-100EA 10900 NOK
ABCAAB62666-100
Anti-D-glucosamine Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: D-glucosamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå