Anti-PIK3R5 Rabbit polyclonal antibody unconjugated

ABCAAB62792-400EA 7725 NOK
ABCAAB62792-400
Anti-PIK3R5 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: PIK3R5 (phosphoinositide-3-kinase, regulatory subunit 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR