Anti-ITGAM Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB62817-200EA 11725 NOK
ABCAAB62817-200
Anti-ITGAM Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR