Anti-APH1A Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB62821-200EA 8425 NOK
ABCAAB62821-200
Anti-APH1A Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: APH1A (APH1A gamma secretase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR