Anti-FAU Rabbit polyclonal antibody unconjugated

ABCAAB63066-400EA 7775 NOK
ABCAAB63066-400
Anti-FAU Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: FAU (Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR