Anti-FIGF Rabbit polyclonal antibody unconjugated

ABCAAB63068-400EA 7600 NOK
ABCAAB63068-400
Anti-FIGF Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: FIGF (c-fos induced growth factor (vascular endothelial growth factor D))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR