Anti-SOST Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63097-400EA 8925 NOK
ABCAAB63097-400
Anti-SOST Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SOST (sclerostin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR