Anti-CCR5 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63123-50EA 8350 NOK
ABCAAB63123-50
Anti-CCR5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR