Anti-LDLRAP1 Rabbit polyclonal antibody unconjugated

ABCAAB63155-400EA 6825 NOK
ABCAAB63155-400
Anti-LDLRAP1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: LDLRAP1 (low density lipoprotein receptor adaptor protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR