Anti-MOGAT2 Rabbit polyclonal antibody unconjugated

ABCAAB63156-400EA 7175 NOK
ABCAAB63156-400
Anti-MOGAT2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: MOGAT2 (monoacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR