Anti-PYGM Rabbit polyclonal antibody unconjugated

ABCAAB63158-400EA 7725 NOK
ABCAAB63158-400
Anti-PYGM Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: PYGM (phosphorylase, glycogen, muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR