Anti-HIF1AN Rabbit polyclonal antibody unconjugated

ABCAAB63163-400EA 7600 NOK
ABCAAB63163-400
Anti-HIF1AN Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: HIF1AN (hypoxia inducible factor 1, alpha subunit inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR