Anti-IRGM Mouse monoclonal antibody unconjugated [clone: 403CT8.5.5]

ABCAAB63230-200EA 7600 NOK
ABCAAB63230-200
Anti-IRGM Mouse monoclonal antibody unconjugated [clone: 403CT8.5.5]
Type: Primary
Antigen: IRGM (immunity-related GTPase family, M)
Clonality: monoclonal
Clone: 403CT8.5.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR