Anti-Melanoma gp100 Mouse Monoclonal Antibody [clone: 3F272]

Leverandør: Abcam
ABCAAB63297-100EA 10925 NOK
ABCAAB63297-100
Anti-Melanoma gp100 Mouse Monoclonal Antibody [clone: 3F272]
Antistoffer
Type: Primary
Antigen: Melanoma gp100
Clonality: monoclonal
Clone: 3F272
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR