Anti-TFRC Rat Monoclonal Antibody (Biotin) [clone: 1.BB.713]
Leverandør: Abcam
ABCAAB63333-100EA 11400 NOK
ABCAAB63333-100
Anti-TFRC Rat Monoclonal Antibody (Biotin) [clone: 1.BB.713]
Antistoffer
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone: 1.BB.713
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse