Anti-DDR2 Mouse monoclonal antibody unconjugated [clone: 3B11E4]

Leverandør: Abcam
ABCAAB63337-100EA 12100 NOK
ABCAAB63337-100
Anti-DDR2 Mouse monoclonal antibody unconjugated [clone: 3B11E4]
Antistoffer
Type: Primary
Antigen: DDR2 (discoidin domain receptor tyrosine kinase 2)
Clonality: monoclonal
Clone: 3B11E4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR