Anti-SMAD1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB63356-100EA 12100 NOK
ABCAAB63356-100
Anti-SMAD1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse