Anti-DDIT3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB63392-100EA 11475 NOK
ABCAAB63392-100
Anti-DDIT3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR