Anti-Serum Albumin Chicken Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Leverandør: Abcam
ABCAAB63498-500EA 11400 NOK
ABCAAB63498-500
Anti-Serum Albumin Chicken Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Primary
Antigen: Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human