Anti-Protein L Chicken Polyclonal Antibody

Leverandør: Abcam
ABCAAB63503-250EA 8350 NOK
ABCAAB63503-250
Anti-Protein L Chicken Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Protein L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR