Anti-Protein L Chicken Polyclonal Antibody (Biotin)

Leverandør: Abcam
ABCAAB63505-250EA 8350 NOK
ABCAAB63505-250
Anti-Protein L Chicken Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Protein L
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR