Anti-MTOR Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB63552-100EA 11475 NOK
ABCAAB63552-100
Anti-MTOR Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MTOR (mechanistic target of rapamycin (serine/threonine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat