Anti-NCF1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB63554-100EA 11475 NOK
ABCAAB63554-100
Anti-NCF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NCF1 (neutrophil cytosolic factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human