Anti-LIN28A Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63740-100EA 11925 NOK
ABCAAB63740-100
Anti-LIN28A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LIN28A (lin-28 homolog A (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR