Anti-RAD51 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63798-100EA 8350 NOK
ABCAAB63798-100
Anti-RAD51 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR