Anti-LC3B Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB63817-100EA 12900 NOK
ABCAAB63817-100
Anti-LC3B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LC3B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat