Anti-GJA1 Rabbit polyclonal antibody unconjugated

ABCAAB63851-100EA 7775 NOK
ABCAAB63851-100
Anti-GJA1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: GJA1 (gap junction protein, alpha 1, 43kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR