Anti-MMP3 Rabbit polyclonal antibody unconjugated

ABCAAB63853-100EA 7775 NOK
ABCAAB63853-100
Anti-MMP3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR