Vårt Oslo-kontor flytter den 25. mars 2019 til Brynsalleen 4, 0667 Oslo
Anti-LILRB3 Mouse monoclonal antibody FITC (Fluorescein) [clone: 4F9]
Leverandør: Abcam
Type: Primary
Antigen: LILRB3 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3)
Clonality: monoclonal
Clone: 4F9
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
100 µg 50 µG ABCAAB63858-50
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen symbol LILRB3
Antigen name leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3
Reaktivitet Human
Konjugasjon FITC (Fluorescein Isothiocyanate)
Klonalitet Monoclonal
antistofftype Primary
Klon 4F9
Vert Mouse
Isotype IgG1
Flow cytometry Yes
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall