Anti-SULT1E1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63877-100EA 8450 NOK
ABCAAB63877-100
Anti-SULT1E1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SULT1E1 (sulfotransferase family 1E, estrogen-preferring, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR