Anti-HIV1 Nef Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB63918-100EA 11475 NOK
ABCAAB63918-100
Anti-HIV1 Nef Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: HIV1 Nef
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå