Anti-BACE1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB63954-100EA 10925 NOK
ABCAAB63954-100
Anti-BACE1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BACE1 (beta-site APP-cleaving enzyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR