Anti-Thyroid Hormone Receptor Goat Polyclonal Antibody

Leverandør: Abcam
ABCAAB63962-200EA 8425 NOK
ABCAAB63962-200
Anti-Thyroid Hormone Receptor Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Thyroid Hormone Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR