Anti-HIST2H2BE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

ABCAAB64039-100EA 8775 NOK
ABCAAB64039-100
Anti-HIST2H2BE Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Type: Primary
Antigen: HIST2H2BE (histone cluster 2, H2be)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR