Anti-TFG2 Rabbit polyclonal antibody unconjugated

ABCAAB64067-100EA 7175 NOK
ABCAAB64067-100
Anti-TFG2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: TFG2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR